Please login to your online account to display your discounted pricing

anti-Annexin A1, Clone: 2F1, Novus Biologicals™

$139.00 - $362.00

Specifications

Clone 2F1
Host Species Mouse
Isotype IgG1
Immunogen Partial recombinant human Annexin A1 (between residues 100-300) expressed in &italic_begin;E. coli&italic_end; [UniProt P04083]
Antigen Annexin A1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223485

 
novus biologicals canada
NBP223485
Unlabeled 0.1mL Each for $362.00

NBP223485SS

 
novus biologicals canada
NBP223485SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 2F1
Host Species Mouse
Isotype IgG1
Immunogen Partial recombinant human Annexin A1 (between residues 100-300) expressed in &italic_begin;E. coli&italic_end; [UniProt P04083]
Antigen Annexin A1
Monoclonal or Polyclonal Monoclonal
Format Precipitation
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. P04083
Gene ID (Entrez) 301
Formulation PBS, 0.5% protein stabilizer
Concentration 1.0mg/mL