Please login to your online account to display your discounted pricing

anti-Angiopoietin-1, Clone: MM0019-26B10, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11085464SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11085464T

  • / Each
 

Description & Specifications

Specifications

Reconstitution Reconstitute the antibody with 500μL sterile 1X PBS (or with 40μL H2O plus 460μL PBS), to give a final concentration of 200μg/mL. Alternatively, reconstitute the antibody in 40μL H2O if a small volume is preferred.
Cross Reactivity Chicken
Cross Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Purified
Host Species Mouse
Clone MM0019-26B10
Antigen Angiopoietin-1
Immunogen purified human recombinant human Angiopoietin-1.
Quantity 0.02mg
Isotype IgG2
Regulatory Status RUO
Gene ID (Entrez) 284
Formulation Lyophilized from a 0.2μm filtered solution in phosphate-buffered saline (PBS).
Gene Accession No. Q15389
Monoclonal or Polyclonal Monoclonal
Test Specificity This recognizes human Ang1 without cross reactivity with human Ang2. Chicken data from customer review.
Conjugate Unlabeled
Primary or Secondary Primary