Please login to your online account to display your discounted pricing

anti-Androgen R/NR3C4 (p Ser213, p Ser210), Clone: 156C135.2, Novus Biologicals™

$139.00 - $373.00

Specifications

Clone 156C135.2
Host Species Mouse
Isotype κ
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 207-221 (GRAREAS*GAPTSSKD) of human androgen receptor, containing the serine 213 phosphorylation site: GenBank Accession No. A39248. Note: S* refers to phosphorylated serine in the peptide sequence.
Antigen Androgen R/NR3C4 (p Ser213
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056603

 
novus biologicals canada
NB10056603
Unlabeled 0.1mg Each for $373.00

NB10056603T

 
novus biologicals canada
NB10056603SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 156C135.2
Host Species Mouse
Isotype κ
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 207-221 (GRAREAS*GAPTSSKD) of human androgen receptor, containing the serine 213 phosphorylation site: GenBank Accession No. A39248. Note: S* refers to phosphorylated serine in the peptide sequence.
Antigen Androgen R/NR3C4 (p Ser213
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity In IGF-1 stimulated LNCaP cells (passage number 38), a ∽110 kDa band was observed. Please see Lin et al. 2003 for additional details. The serine phosphorylation site recognized by this antibody has been alternatively referred to Ser213 (Lee and Chang, 2003) and Ser210 (Lin et al, 2003). Variations in denotation can arise from how the sequence is counted in various GenBank accession numbers. The site is denoted as Ser213 in GenBank Accession No. A39248, which was used to design the immunogen. Human and New World Monkey. Predicted cross-reactivity with Chimpanzee, Dog, Mouse, Pig and Rat.
Gene Accession No. P10275
Gene ID (Entrez) 367
Formulation 0.2mL PBS containing 0.05% BSA
Concentration 0.5mg/mL