Please login to your online account to display your discounted pricing

anti-alpha-Fetoprotein/AFP, Clone: C3, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone C3
Host Species Mouse
Isotype κ
Immunogen Alpha fetoprotein (AFP) purified from serum of a hepatoma patient
Antigen alpha-Fetoprotein/AFP
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329141M

 
novus biologicals canada
NBP2329141ML
Unlabeled 1mL Each for $429.00

NBP232914

 
novus biologicals canada
NBP232914
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone C3
Host Species Mouse
Isotype κ
Immunogen Alpha fetoprotein (AFP) purified from serum of a hepatoma patient
Antigen alpha-Fetoprotein/AFP
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Bovine (Negative), Feline (Negative), Mouse (Negative), Rat (Negative).
Gene Accession No. P02771
Gene ID (Entrez) 174
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL