Please login to your online account to display your discounted pricing

anti-alpha-Fetoprotein/AFP, Clone: C3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329141ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329141M

  • / Each
 

Description & Specifications

Specifications

Antigen alpha-Fetoprotein/AFP
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone C3
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Primate
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P02771
Host Species Mouse
Immunogen Alpha fetoprotein (AFP) purified from serum of a hepatoma patient
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 174
Test Specificity Bovine (Negative), Feline (Negative), Mouse (Negative), Rat (Negative).