Please login to your online account to display your discounted pricing

anti-Aldolase C, Clone: 1A1, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 1A1
Host Species Murine
Isotype IgG1
Immunogen The C-terminal 23 amino acids of human Aldolase C (KYEGSGEDGGAAAQSLYIANHAY). [UniProt No. P09972]
Antigen Aldolase C
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225144

 
novus biologicals canada
NBP225144
Unlabeled 0.1mL Each for $309.05

NBP225144SS

 
novus biologicals canada
NBP225144SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone 1A1
Host Species Murine
Isotype IgG1
Immunogen The C-terminal 23 amino acids of human Aldolase C (KYEGSGEDGGAAAQSLYIANHAY). [UniProt No. P09972]
Antigen Aldolase C
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Rat. Other species have not been tested.
Gene ID (Entrez) 230
Formulation PBS
Concentration 1mg/mL