Please login to your online account to display your discounted pricing

anti-Akt3, Clone: 66C1247, Novus Biologicals™

$139.00 - $307.00

Specifications

Clone 66C1247
Host Species Mouse
Isotype IgG1
Immunogen GenBank no. gi4574744gbAAD24196.1AF135794_1. This sequence is identical in human
Antigen Akt3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001298

 
novus biologicals canada
NB6001298
Unlabeled 0.1mg Each for $307.00

NB6001298SS

 
novus biologicals canada
NB6001298SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 66C1247
Host Species Mouse
Isotype IgG1
Immunogen GenBank no. gi4574744gbAAD24196.1AF135794_1. This sequence is identical in human
Antigen Akt3
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Cross-reacts with Human, Mouse and Rat. Not yet tested in other species.
Gene Accession No. Q9Y243
Gene ID (Entrez) 10000
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL