Please login to your online account to display your discounted pricing

anti-Akt2, Clone: 95C657.1.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056676SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056676T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P31751
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Format Ascites
Formulation PBS containing 0.5% BSA
Regulatory Status RUO
Gene ID (Entrez) 208
Test Specificity AKT2 (95C657.1.2).
Cross Reactivity Human
Antigen Akt2
Applications Western Blotting
Immunogen This antibody was generated by immunizing mice with synthetic peptide VANSLKQRAPGEDPMDYK (amino acids 106-123) of human AKT2.
Isotype IgM
Quantity 0.05mL
Clone 95C657.1.2
Host Species Mouse