Please login to your online account to display your discounted pricing

anti-AIP/ARA9, Clone: 35-2, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100127SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100127SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene Accession No. O00170
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Primary or Secondary Primary
Test Specificity Customers have reported success on rat lysate (see review).
Gene ID (Entrez) 9049
Monoclonal or Polyclonal Monoclonal
Antigen AIP/ARA9
Isotype IgG1
Applications Chromatin Immunoprecipitation
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Regulatory Status RUO
Quantity 0.025mL
Format Supernatant
Immunogen Bacterially expressed human AIP/ARA9 [UniProt No. O00170]
Host Species Mouse
Clone 35-2