Please login to your online account to display your discounted pricing

anti-Actin Alpha 2 Smooth Muscle, Clone: 1A4/asm-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233006

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233006

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Concentration 0.2mg/mL
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P62736
Gene ID (Entrez) 59
Conjugate Unlabeled
Quantity 0.5mL
Cross Reactivity Baboon
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Goat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Format Purified
Regulatory Status RUO
Antigen Actin Alpha 2 Smooth Muscle
Host Species Mouse
Immunogen N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH
Clone 1A4/asm-1