Please login to your online account to display your discounted pricing

anti-Actin Alpha 2 Smooth Muscle, Clone: 1A4/asm-1, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone 1A4/asm-1
Host Species Mouse
Isotype κ
Immunogen N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH
Antigen Actin Alpha 2 Smooth Muscle
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP233006

 
novus biologicals canada
NBP233006
Unlabeled 0.5mL Each for $321.00

NBP2330061M

 
novus biologicals canada
NBP2330061ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone 1A4/asm-1
Host Species Mouse
Isotype κ
Immunogen N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH
Antigen Actin Alpha 2 Smooth Muscle
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P62736
Gene ID (Entrez) 59
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL