Please login to your online account to display your discounted pricing

anti-Actin Alpha 2 Smooth Muscle, Clone: 1A4/asm-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2330061ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2330061M

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Conjugate Unlabeled
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P62736
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Gene ID (Entrez) 59
Cross Reactivity Baboon
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Goat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Quantity 1mL
Regulatory Status RUO
Clone 1A4/asm-1
Immunogen N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH
Isotype κ
Antigen Actin Alpha 2 Smooth Muscle
Host Species Mouse