Please login to your online account to display your discounted pricing

anti-ACTH, Clone: 57, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329121ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329121M

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene Accession No. P01189
Gene ID (Entrez) 5443
Concentration 0.2mg/mL
Test Specificity Expected to show broad species reactivity.
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Isotype κ
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Clone 57
Format Purified
Quantity 1mL
Antigen ACTH
Immunogen N-terminal fragment of human ACTH conjugated to KLH
Regulatory Status RUO