Please login to your online account to display your discounted pricing

anti-ACTH, Clone: 57, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232912

 View all options for this product


For Research Use Only

Catalog No. NBP232912


Description & Specifications


Antigen ACTH
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone 57
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01189
Host Species Mouse
Immunogen N-terminal fragment of human ACTH conjugated to KLH
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5443
Test Specificity Expected to show broad species reactivity.