Please login to your online account to display your discounted pricing

anti-ACTH, Clone: 57, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone 57
Host Species Mouse
Isotype κ
Immunogen N-terminal fragment of human ACTH conjugated to KLH
Antigen ACTH
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232912

 
novus biologicals canada
NBP232912
Unlabeled 0.5mL Each for $321.00

NBP2329121M

 
novus biologicals canada
NBP2329121ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone 57
Host Species Mouse
Isotype κ
Immunogen N-terminal fragment of human ACTH conjugated to KLH
Antigen ACTH
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Expected to show broad species reactivity.
Gene Accession No. P01189
Gene ID (Entrez) 5443
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL