Please login to your online account to display your discounted pricing

anti-ABCG8, Clone: 1B10A5, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 1B10A5
Host Species Mouse
Isotype IgG
Immunogen Full length mouse ABCG8 [Swiss-Prot No. Q9DBM0]
Antigen ABCG8
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP171706

 
novus biologicals canada
NBP171706
Unlabeled 0.1mL Each for $329.00

NBP171706SS

 
novus biologicals canada
NBP171706SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 1B10A5
Host Species Mouse
Isotype IgG
Immunogen Full length mouse ABCG8 [Swiss-Prot No. Q9DBM0]
Antigen ABCG8
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 64241
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL