Please login to your online account to display your discounted pricing

anti-ABCG2/CD338, Clone: 3G8, Novus Biologicals™

$144.63 - $362.00

Specifications

Clone 3G8
Host Species Mouse
Isotype IgG1
Immunogen Purified recombinant fragment of human ABCG2 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. Q9UNQ0]
Antigen ABCG2/CD338
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222124

 
novus biologicals canada
NBP222124
Unlabeled 0.1mL Each for $362.00

NBP222124SS

 
novus biologicals canada
NBP222124SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Clone 3G8
Host Species Mouse
Isotype IgG1
Immunogen Purified recombinant fragment of human ABCG2 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. Q9UNQ0]
Antigen ABCG2/CD338
Monoclonal or Polyclonal Monoclonal
Format Precipitation
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. Q9UNQ0
Gene ID (Entrez) 9429
Formulation PBS
Concentration 1.0mg/mL