Please login to your online account to display your discounted pricing

anti-ABCA1, Clone: HJ1, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone HJ1
Host Species Murine
Isotype κ
Immunogen 50 kDa N-terminal extracellular loop of mouse ABCA1 [UniProt No. P41233]
Antigen ABCA1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002068SS

 
novus biologicals canada
NB1002068SS
Unlabeled 0.025mL Each for $139.00

NB1002068

 
novus biologicals canada
NB1002068
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Clone HJ1
Host Species Murine
Isotype κ
Immunogen 50 kDa N-terminal extracellular loop of mouse ABCA1 [UniProt No. P41233]
Antigen ABCA1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene ID (Entrez) 19
Formulation PBS containing protease inhibitors
Concentration 1mg/mL