Please login to your online account to display your discounted pricing

anti-A2BP1, Clone: 1G10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225141

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225141

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Test Specificity Human, Bovine, Mouse and Rat. Other species have not been tested.
Primary or Secondary Primary
Gene ID (Entrez) 54715
Conjugate Unlabeled
Regulatory Status RUO
Immunogen The N-terminal 100 amino acids of human A2BP1 expressed in and purified from &italic_begin;E. coli&italic_end;. [UniProt No. Q9NWB1]
Isotype κ
Quantity 0.1mL
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Antigen A2BP1
Clone 1G10
Gene Accession No. Q9NWB1
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL