Please login to your online account to display your discounted pricing

anti-A2BP1, Clone: 1G10, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 1G10
Host Species Mouse
Isotype κ
Immunogen The N-terminal 100 amino acids of human A2BP1 expressed in and purified from &italic_begin;E. coli&italic_end;. [UniProt No. Q9NWB1]
Antigen A2BP1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225141SS

 
novus biologicals canada
NBP225141SS
Unlabeled 0.025mL Each for $109.00

NBP225141

 
novus biologicals canada
NBP225141
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone 1G10
Host Species Mouse
Isotype κ
Immunogen The N-terminal 100 amino acids of human A2BP1 expressed in and purified from &italic_begin;E. coli&italic_end;. [UniProt No. Q9NWB1]
Antigen A2BP1
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human, Bovine, Mouse and Rat. Other species have not been tested.
Gene Accession No. Q9NWB1
Gene ID (Entrez) 54715
Formulation PBS
Concentration 1mg/mL