Please login to your online account to display your discounted pricing

anti-A2BP1, Clone: 1G10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225141SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225141SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q9NWB1
Immunogen The N-terminal 100 amino acids of human A2BP1 expressed in and purified from &italic_begin;E. coli&italic_end;. [UniProt No. Q9NWB1]
Isotype κ
Quantity 0.025mL
Primary or Secondary Primary
Format Affinity Purified
Regulatory Status RUO
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Antigen A2BP1
Clone 1G10
Host Species Mouse
Gene ID (Entrez) 54715
Test Specificity Human, Bovine, Mouse and Rat. Other species have not been tested.
Formulation PBS
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled