Please login to your online account to display your discounted pricing

anti-A20/TNFAIP3, Clone: 59A426, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177533

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177533

  • / Each
 

Description & Specifications

Specifications

Antigen A20/TNFAIP3
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Clone 59A426
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. P21580
Host Species Murine
Immunogen Full length recombinant human A20. The epitope has been mapped to the C-terminal portion of A20 (amino acids 440-790)
Isotype κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7128
Target Species Human
Target Species Murine
Target Species Rat