Please login to your online account to display your discounted pricing

anti-6-Histidine Epitope Tag, Clone: AD1.1.10, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10064768

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10064768

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL
Test Specificity This antibody is reactive to histidine tags.
Cross Reactivity Non-species specific
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Primary or Secondary Primary
Antigen 6-Histidine Epitope Tag
Isotype IgG1
Clone AD1.1.10
Host Species Mouse
Immunogen PAX6 transcription factor linked to histidine tag
Regulatory Status RUO
Format Purified
Quantity 0.1mL