Please login to your online account to display your discounted pricing

anti-6-Histidine Epitope Tag, Clone: AD1.1.10, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone AD1.1.10
Host Species Mouse
Isotype IgG1
Immunogen PAX6 transcription factor linked to histidine tag
Antigen 6-Histidine Epitope Tag
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10064768T

 
novus biologicals canada
NB10064768SS
Unlabeled 0.025mL Each for $139.00

NB10064768

 
novus biologicals canada
NB10064768
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Clone AD1.1.10
Host Species Mouse
Isotype IgG1
Immunogen PAX6 transcription factor linked to histidine tag
Antigen 6-Histidine Epitope Tag
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Test Specificity This antibody is reactive to histidine tags.
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL