Please login to your online account to display your discounted pricing

anti-5'-Nucleotidase/CD73, Clone: 45M3B4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225235

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225235

  • / Each
 

Description & Specifications

Specifications

Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Gene ID (Entrez) 4907
Gene Accession No. NP_001191742
Primary or Secondary Primary
Concentration 0.5mg/mL
Conjugate Unlabeled
Isotype κ
Antigen 5'-Nucleotidase/CD73
Monoclonal or Polyclonal Monoclonal
Test Specificity This product can detect Isoform 1 and Isoform of Human CD73.
Immunogen Within the range of amino acids 169-324 of human CD73 were used as the immunogen for the antibody.
Regulatory Status RUO
Cross Reactivity Human
Quantity 0.1mg
Host Species Mouse
Format Purified
Applications Western Blotting
Clone 45M3B4