Please login to your online account to display your discounted pricing

anti-5'-Nucleotidase/CD73, Clone: 45M3B4, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 45M3B4
Host Species Mouse
Isotype κ
Immunogen Within the range of amino acids 169-324 of human CD73 were used as the immunogen for the antibody.
Antigen 5'-Nucleotidase/CD73
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225235SS

 
novus biologicals canada
NBP225235SS
Unlabeled 0.025mg Each for $139.00

NBP225235

 
novus biologicals canada
NBP225235
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Clone 45M3B4
Host Species Mouse
Isotype κ
Immunogen Within the range of amino acids 169-324 of human CD73 were used as the immunogen for the antibody.
Antigen 5'-Nucleotidase/CD73
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity This product can detect Isoform 1 and Isoform of Human CD73.
Gene Accession No. NP_001191742
Gene ID (Entrez) 4907
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Concentration 0.5mg/mL