Please login to your online account to display your discounted pricing

anti-5'-Nucleotidase/CD73, Clone: 45M3B4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225235SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225235SS

  • / Each
 

Description & Specifications

Specifications

Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Gene Accession No. NP_001191742
Primary or Secondary Primary
Concentration 0.5mg/mL
Test Specificity This product can detect Isoform 1 and Isoform of Human CD73.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4907
Regulatory Status RUO
Isotype κ
Clone 45M3B4
Conjugate Unlabeled
Cross Reactivity Human
Immunogen Within the range of amino acids 169-324 of human CD73 were used as the immunogen for the antibody.
Quantity 0.025mg
Antigen 5'-Nucleotidase/CD73
Format Purified
Host Species Mouse
Applications Western Blotting