Please login to your online account to display your discounted pricing

anti-3-OHKYN, Clone: P3UI, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100597

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100597

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Test Specificity This is specific for 3-hydroxykynureinine (3OH-Kyn) modifications on proteins. It is likely to react with 3OH-Kyn modification on lysine, histidine and cysteine residues on proteins. It does not react with enzymes involved in the degradation of tryptopha. Human, although there should be no species limitation to this antibody's reactivity.
Concentration 4.3mg/mL
Formulation Tris-glycine, 150mM NaCl
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity All
Cross Reactivity Human
Format Purified
Isotype κ
Regulatory Status RUO
Antigen 3-OHKYN
Host Species Mouse
Quantity 0.1mL
Immunogen 3-OHKYN modified KLH
Clone P3UI