Please login to your online account to display your discounted pricing

anti-3-OHKYN, Clone: P3UI, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone P3UI
Host Species Murine
Isotype κ
Immunogen 3-OHKYN modified KLH
Antigen 3-OHKYN
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100597

 
novus biologicals canada
NB100597
Unlabeled 0.1mL Each for $309.05

NB100597SS

 
novus biologicals canada
NB100597SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone P3UI
Host Species Murine
Isotype κ
Immunogen 3-OHKYN modified KLH
Antigen 3-OHKYN
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity This is specific for 3-hydroxykynureinine (3OH-Kyn) modifications on proteins. It is likely to react with 3OH-Kyn modification on lysine, histidine and cysteine residues on proteins. It does not react with enzymes involved in the degradation of tryptopha. Human, although there should be no species limitation to this antibody's reactivity.
Formulation Tris-glycine, 150mM NaCl
Concentration 4.3mg/mL