Please login to your online account to display your discounted pricing

anti-3-OHKYN, Clone: P3UI, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100597SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100597SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Tris-glycine, 150mM NaCl
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Test Specificity This is specific for 3-hydroxykynureinine (3OH-Kyn) modifications on proteins. It is likely to react with 3OH-Kyn modification on lysine, histidine and cysteine residues on proteins. It does not react with enzymes involved in the degradation of tryptopha. Human, although there should be no species limitation to this antibody's reactivity.
Concentration 4.3mg/mL
Isotype κ
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Purified
Regulatory Status RUO
Cross Reactivity All
Cross Reactivity Human
Immunogen 3-OHKYN modified KLH
Clone P3UI
Host Species Mouse
Antigen 3-OHKYN
Quantity 0.025mL