Please login to your online account to display your discounted pricing

anti-14-3-3 eta, Clone: 3G12, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP192691

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP192691

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Quantity 0.1mL
Antigen 14-3-3 eta
Immunogen Full length recombinant human 14-3-3 eta [UniProt No. Q04917]
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7533
Formulation PBS
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Gene Accession No. Q04917
Primary or Secondary Primary
Host Species Mouse
Isotype IgG1
Regulatory Status RUO
Clone 3G12