Please login to your online account to display your discounted pricing

anti-14-3-3 eta, Clone: 3G12, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 3G12
Host Species Murine
Isotype IgG1
Immunogen Full length recombinant human 14-3-3 eta [UniProt No. Q04917]
Antigen 14-3-3 eta
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP192691SS

 
novus biologicals canada
NBP192691SS
Unlabeled 0.025mL Each for $109.00

NBP192691

 
novus biologicals canada
NBP192691
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone 3G12
Host Species Murine
Isotype IgG1
Immunogen Full length recombinant human 14-3-3 eta [UniProt No. Q04917]
Antigen 14-3-3 eta
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. Q04917
Gene ID (Entrez) 7533
Formulation PBS
Concentration 1mg/mL