Monomethyl-Histone H4 (Lys20) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  Emd Millipore Corporation 071570
 View more versions of this product
Catalog No. 07-157-0MI
Description
Description
Anti-monomethyl Histone H4 (Lys20) Antibody is a rabbit polyclonal antibody for detection of monomethyl Histone H4 (Lys20) also known as H4K20me1, Histone H4 (mono methyl K20), H4 histone family member A and has been validated in western blotting.
Specifications
Specifications
monomethyl Histone H4 (Lys20)
Unconjugated
Affinity Purified
Rabbit
Affinity chromatography
−20°C in undiluted aliquotes for up to 12 months
Polyclonal
Human
ChIP assay,Dot Blot,Inhibition Assays,Western Blot
Purified from serum by affinity chromatography. Supplied at 1mg/mL in 0.1M Tris-Glycine (pH7.4), 15mM NaCl, 0.05% NaN3.
H4/A; H4/B; H4/C; H4/D; H4/E; H4/G ; H4/H; H4/I ; H4/J ; H4/K ; H4/M; H4/N; H4/O; H4/a ; H4F2; H4FA ; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; HIST2H4
KLH-conjugated, linear synthetic peptide corresponding to amino acids 15-24 (AKRHR[me1K]VLRD-C) of human Histone H4.
100μg
Primary
NM_003538.3