Please login to your online account to display your discounted pricing

anti-monomethyl-Histone H3 (Lys9) Ab 2, Polyclonal, EMD Millipore™


Manufacturer: emd millipore corporation  ABE101

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. ABE101MI

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications Immunocytochemistry
Applications Peptide Inhibition Assay
Applications Western Blotting
Gene ID (Entrez) NP_003484.1
Purification Method Affinity Purfied
Format Affinity Purified
Primary or Secondary Prrimary
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
Immunogen KLH-conjugated branched peptide corresponding to human Histone H3 methylated at Lys9.
Antigen monomethyl-Histone H3 (Lys9)
Monoclonal or Polyclonal Polyclonal
Storage Requirements 2-8°C for 12 months
Host Species Rabbit
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Conjugate Unlabeled
Quantity 100μg