missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Monomethyl-Histone H3 (Lys9) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$127.00 - $506.00

Specifications

Host Species Rabbit
Immunogen KLH-conjugated branched peptide corresponding to human Histone H3 methylated at Lys9.
Antigen monomethyl-Histone H3 (Lys9)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ABE101SMI View Documents Emd Millipore Corporation
ABE101S
10μg Each for $127.00
 
ABE101MI View Documents Emd Millipore Corporation
ABE101
100μg Each for $506.00
 
Description

Description

Anti-monomethyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 monomethylated on lysine 9. This purified Ab, also known as Anti-H3K9me1 has dot blot proven specificity and is validated in western blot, immunocytochemistry, PIA.
Specifications

Specifications

Rabbit
monomethyl-Histone H3 (Lys9)
Affinity Purified
Immunocytochemistry,Inhibition Assays,Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
KLH-conjugated branched peptide corresponding to human Histone H3 methylated at Lys9.
Polyclonal
Primary
NP_003484.1

For Research Use Only