missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Monomethyl-Histone H3 (Lys9) Rabbit anti-Human, Mouse, Chicken, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer: Emd Millipore Corporation 07450
Description
Anti-monomethyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 monomethylated on lysine 9 (H3K9me1). This purified antibody is specificity verified by dot blot (DB), published in peer reviewed journals, and has been validated in immunocytochemistry, PIA and western blotting.Specifications
monomethyl-Histone H3 (Lys9) | |
Unconjugated | |
Purified | |
Rabbit | |
IgG | |
100μg | |
Primary | |
NM_003493 |
Dot Blot,Immunocytochemistry,Inhibition Assays,Western Blot | |
Purified rabbit IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol. | |
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886 | |
KLH-conjugated | |
Protein A purfied | |
−20°C in undiluted aliquotes for up to 12 months | |
Polyclonal | |
Chicken,Human,Murine |
For Research Use Only