missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Monomethyl-Histone H3 (Lys36) Rabbit anti-Mouse, Human, Rat, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$118.34 - $474.33

Specifications

Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H3 monomethylated at Lys36.
Antigen monomethyl-Histone H3 (Lys36)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ABE244SMI View Documents MilliporeSigma
ABE244S
10μg Each for $118.34
 
ABE244MI View Documents MilliporeSigma
ABE244
100μg Each for $474.33
 
Description

Description

Anti-monomethyl-Histone H3 (Lys36) Antibody is a rabbit polyclonal antibody for detection of Histone H3 monomethylated at lysine 36. This purified polyclonal antibody also known as Anti-H3K36me1, has dot blot (DB) proven specificity and has been validated in western blot, immunoprecipitation.
Specifications

Specifications

Rabbit
monomethyl-Histone H3 (Lys36)
Affinity Purified
Dot Blot, Immunoprecipitation, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H3 monomethylated at Lys36.
Polyclonal
Primary
NP_003484

For Research Use Only