Monomethyl-Histone H3 (Lys36) Rabbit anti-Mouse, Human, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE244S
Description
Anti-monomethyl-Histone H3 (Lys36) Antibody is a rabbit polyclonal antibody for detection of Histone H3 monomethylated at lysine 36. This purified polyclonal antibody also known as Anti-H3K36me1, has dot blot (DB) proven specificity and has been validated in western blot, immunoprecipitation.Specifications
monomethyl-Histone H3 (Lys36) | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Dot Blot, Immunoprecipitation, Western Blot | |
Unconjugated | |
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886 | |
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H3 monomethylated at Lys36. | |
10 μg | |
NP_003484 | |
2-8°C for 12 months |
For Research Use Only