Please login to your online account to display your discounted pricing

anti-MMP-7, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human MMP7 protein (between residues 250-300) [UniProt P09237]
Antigen MMP-7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP199123SS

 
novus biologicals canada
NBP199123SS
Unlabeled 0.025mL Each for $139.00

NBP199123

 
novus biologicals canada
NBP199123
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human MMP7 protein (between residues 250-300) [UniProt P09237]
Antigen MMP-7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Gene Accession No. P09237
Gene ID (Entrez) 4316
Formulation PBS (pH 7.4)
Concentration 1.28mg/mL