Please login to your online account to display your discounted pricing

anti-MMP-2, Polyclonal, Novus Biologicals™

$144.63 - $384.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human MMP2 protein (between residues 400-500) [UniProt P08253]
Antigen MMP-2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB200193

 
novus biologicals canada
NB200193
Unlabeled 0.1mL Each for $384.00

NB200193SS

 
novus biologicals canada
NB200193SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human MMP2 protein (between residues 400-500) [UniProt P08253]
Antigen MMP-2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity This antibody is specific for pro and active forms of MMP2. It does not react with other MMPs tested.
Gene Accession No. P08253
Gene ID (Entrez) 4313
Formulation PBS
Concentration 1mg/mL