Please login to your online account to display your discounted pricing

anti-MMP-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB200193SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB200193SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Gene Accession No. P08253
Gene ID (Entrez) 4313
Conjugate Unlabeled
Primary or Secondary Primary
Test Specificity This antibody is specific for pro and active forms of MMP2. It does not react with other MMPs tested.
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Host Species Rabbit
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Affinity Purified
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen MMP-2
Immunogen A synthetic peptide made to an internal region of the human MMP2 protein (between residues 400-500) [UniProt P08253]
Quantity 0.025mL