Please login to your online account to display your discounted pricing

anti-MMP-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB200193

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB200193

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Conjugate Unlabeled
Formulation PBS
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Gene Accession No. P08253
Gene ID (Entrez) 4313
Test Specificity This antibody is specific for pro and active forms of MMP2. It does not react with other MMPs tested.
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen MMP-2
Quantity 0.1mL
Format Affinity Purified
Regulatory Status RUO
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human MMP2 protein (between residues 400-500) [UniProt P08253]