Please login to your online account to display your discounted pricing

anti-MMP-1, Polyclonal, Novus Biologicals™

$142.00 - $445.44

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-term of human MMP-1.
Antigen MMP-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP172209

 
novus biologicals canada
NBP172209
Unlabeled 0.1mg Each for $445.44

NBP172209SS

 
novus biologicals canada
NBP172209SS
Unlabeled 0.02mg Each for $142.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-term of human MMP-1.
Antigen MMP-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This antibody recognizes human MMP-1 (Interstitial collagenase/ gelatinase A) with both non-cleaved (50kDa) and cleaved forms of (22kDa and 27 kDa). The other species are not tested.
Gene Accession No. P03956
Gene ID (Entrez) 4312
Formulation PBS (pH 7.2) containing antibody stabilizer.
Concentration 0.5mg/mL