Please login to your online account to display your discounted pricing

anti-MMP-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP172209

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP172209

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4312
Primary or Secondary Primary
Test Specificity This antibody recognizes human MMP-1 (Interstitial collagenase/ gelatinase A) with both non-cleaved (50kDa) and cleaved forms of (22kDa and 27 kDa). The other species are not tested.
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Formulation PBS (pH 7.2) containing antibody stabilizer.
Gene Accession No. P03956
Immunogen A synthetic peptide corresponding to C-term of human MMP-1.
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Quantity 0.1mg
Antigen MMP-1
Host Species Rabbit
Regulatory Status RUO