Please login to your online account to display your discounted pricing

anti-Mitofusin 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP151841SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP151841SS

  • / Each
 

Description & Specifications

Specifications

Antigen Mitofusin 1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 1.15mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS
Gene Accession No. Q8IWA4
Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of human Mitofusin-1 (within residues 5-100). [Swiss-Prot Q8IWA4]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 55669
Target Species Human
Target Species Murine
Target Species Rat