Please login to your online account to display your discounted pricing

anti-MICB, PerCP, Clone: 236511, R&D Systems


Manufacturer: r & d systems  RDSFAB1599C100

 View more versions of this product

Catalog No. FAB1599C100

  • / Each
 

Description & Specifications

Specifications

Immunogen Mouse myeloma cell line NS0-derived recombinant human MICB. Ala23-Gly298
Isotype IgG2b
Cross Reactivity Human
Format Purified
Antigen MICB
Quantity 100 Tests
Primary or Secondary Primary
Conjugate PerCP
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Clone 236511
Purification Method Protein A or G purified from hybridoma culture supernatant
Regulatory Status RUO