Please login to your online account to display your discounted pricing

anti-MICB, PerCP, Clone: 236511, R&D Systems


Manufacturer: r & d systems  RDSFAB1599C025

 View more versions of this product

Catalog No. FAB1599C025

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Isotype IgG2b
Host Species Mouse
Clone 236511
Format Purified
Quantity 25 Tests
Conjugate PerCP
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Antigen MICB
Immunogen Mouse myeloma cell line NS0-derived recombinant human MICB. Ala23-Gly298
Purification Method Protein A or G purified from hybridoma culture supernatant
Cross Reactivity Human