Please login to your online account to display your discounted pricing

anti-MICB, PerCP, Clone: 236511, R&D Systems

$177.37 - $445.33

Specifications

Antigen MICB
Clone 236511
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB1599C100

 
r & d systems
RDSFAB1599C100
PerCP 100 Tests Each for $445.33

FAB1599C025

 
r & d systems
RDSFAB1599C025
PerCP 25 Tests Each for $177.37
Description & Specifications

Specifications

Antigen MICB
Clone 236511
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant human MICB. Ala23-Gly298
Isotype IgG2b
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry