Please login to your online account to display your discounted pricing

anti-MICB, PE, Clone: 236511, R&D Systems


Manufacturer: r & d systems  RDSFAB1599P100

 View more versions of this product

Catalog No. FAB1599P100

  • / Each
 

Description & Specifications

Specifications

Isotype IgG2b
Host Species Mouse
Format Purified
Clone 236511
Purification Method Protein A or G purified from hybridoma culture supernatant
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate PE
Regulatory Status RUO
Quantity 100 Tests
Antigen MICB
Immunogen Mouse myeloma cell line NS0-derived recombinant human MICB. Ala23-Gly298
Cross Reactivity Human