Please login to your online account to display your discounted pricing

anti-MICB, APC, Clone: 236511, R&D Systems


Manufacturer: r & d systems  RDSFAB1599A100

 View more versions of this product

Catalog No. FAB1599A100

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Cross Reactivity Human
Clone 236511
Quantity 100 Tests
Immunogen Mouse myeloma cell line NS0-derived recombinant human MICB. Ala23-Gly298
Format Purified
Conjugate APC
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Isotype IgG2b
Antigen MICB
Host Species Mouse
Purification Method Protein A or G purified from hybridoma culture supernatant